missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p120-catenin Antibody (OTI2E8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73179B
This item is not returnable.
View return policy
Description
p120-catenin Monoclonal specifically detects p120-catenin in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| p120-catenin | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CTNND1 | |
| Full length recombinant protein of human p120-catenin (NP_001078927) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2E8 | |
| Western Blot, Immunohistochemistry | |
| Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific, Signal Transduction | |
| 1500.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction