missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p120 Catenin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12180-1-AP-20UL
This item is not returnable.
View return policy
Description
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.Specifications
| p120 Catenin | |
| Polyclonal | |
| Unconjugated | |
| CTNND1 | |
| CAS, Catenin delta 1, CTNND, CTNND1, KIAA0384, p120, p120 Catenin, p120(cas), p120(ctn), P120CAS, p120-catenin, P120CTN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12388, 1500, 311163 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60716, P30999 | |
| Ctnnd1 | |
| p120 Catenin Fusion Protein Ag2824 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction