missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p130 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12776-100ul
This item is not returnable.
View return policy
Description
p130 Polyclonal antibody specifically detects p130 in Human samples. It is validated for Western Blot
Specifications
| p130 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| FLJ26459, P130, PRB2, RB2, RBR-2, retinoblastoma-like 2 (p130), retinoblastoma-like protein 2,130 kDa retinoblastoma-associated protein, Retinoblastoma-related protein 2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human p130 | |
| 100 μg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 5934 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction