missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p130Cas/BCAR1 Antibody (SR1123), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | p130Cas/BCAR1 |
|---|---|
| Clone | SR1123 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694355
|
Novus Biologicals
NBP3-22181-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624324
|
Novus Biologicals
NBP3-22181-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p130Cas/BCAR1 Monoclonal antibody specifically detects p130Cas/BCAR1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| p130Cas/BCAR1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas | |
| A synthesized peptide derived from human p130Cas/BCAR1 (Uniprot #: P56945) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1123 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9564 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title