missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p130Cas Monoclonal Antibody (M144)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547675
This item is not returnable.
View return policy
Description
The antibody detects a 130 kDa* protein corresponding to the molecular mass of p130 Cas on SDS-PAGE immunoblots of human A431, endothelial, and Hct116 cells.
The p130Cas (Cas for Crk-associated substrate) is a common cellular target of phosphorylation signal via v-Crk and v-Src oncoproteins. p130Cas has a unique structure that contains a Src homology (SH)-3 domain followed by multiple YXXP motifs and a proline-rich regionp130Cas is implicated in a variety of biological processes including cell adhesion, cell migration, growth factor stimulation, cytokine receptor engagement and bacterial infection. Physiological functions of p130Cas include cardiovascular development, actin filament assembly and Src-induced cell transformation.
Specifications
| p130Cas | |
| Monoclonal | |
| 0.0625 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P56945, Q61140, Q63767 | |
| BCAR1 | |
| Clone M144 was generated from a recombinant protein containing amino acid residues in the C-terminal region of rat p130 Cas. This rat sequence is highly conserved in human, mouse, and chicken p130 Cas. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| M144 | |
| Unconjugated | |
| BCAR1 | |
| AI385681; BCA1; Bcar1; BCAR1 scaffold protein, Cas family member; BCAR1, Cas family scaffold protein; BCAR1, Cas family scaffolding protein; breast cancer anti-estrogen resistance 1; breast cancer anti-estrogen resistance protein 1; Cas; Cas scaffolding protein family member 1; CAS1; CASS1; CRKAS; CRK-associated substrate; Crk-associated substrate p130Cas; OTTHUMP00000174941; p130 Cas; P130CAS; p130Cas (pTyr410); p130Cas (pY410); p130Cas phospho; phospho p130Cas; v-crk-associated tyrosine kinase substrate | |
| Mouse | |
| Protein A | |
| RUO | |
| 12927, 25414, 9564 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction