missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p16INK4a/CDKN2A Antibody (CDKN2A/8196R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20901CL1
This item is not returnable.
View return policy
Description
p16INK4a/CDKN2A Monoclonal antibody specifically detects p16INK4a/CDKN2A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| p16INK4a/CDKN2A | |
| Monoclonal | |
| CoraFluor 1 | |
| ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
| Purified recombinant prokaryotic full-length human p16INK4a/CDKN2A protein | |
| 0.1 mL | |
| ABC Transporters, Biologically Active Proteins, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, Tumor Suppressors | |
| 1029 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| CDKN2A/8196R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction