Learn More
p16INK4a/CDKN2A Antibody (CDKN2A/8223R), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20902V
Product Code. 18542869
  • € 726.00 / 0.10mL
Description
p16INK4a/CDKN2A Monoclonal antibody specifically detects p16INK4a/CDKN2A in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p16INK4a/CDKN2A
Monoclonal
DyLight 405
ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16
Purified recombinant prokaryotic full-length human p16INK4a/CDKN2A protein
0.1 mL
ABC Transporters, Biologically Active Proteins, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, Tumor Suppressors
1029
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
CDKN2A/8223R
50 mM Sodium Borate
Rabbit
Protein A or G purified