missing translation for 'onlineSavingsMsg'
Learn More

p18INK4c/CDKN2C Antibody (OTI2E1), PE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-73185PE

Product Code. 30029383

  • € 740.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

p18INK4c/CDKN2C Monoclonal antibody specifically detects p18INK4c/CDKN2C in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

p18INK4c/CDKN2C
Monoclonal
PE
PBS
Mouse
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
OTI2E1
Western Blot
CDK6 inhibitor p18, CDKN6, cyclin-dependent inhibitor, cyclin-dependent kinase 4 inhibitor C, Cyclin-dependent kinase 6 inhibitor, cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4), INK4Ccyclin-dependent kinase 6 inhibitor p18, p18, p18-INK4C, p18-INK6
Full length human recombinant protein of human CDKN2C (NP_001253) produced in E.coli.
0.1 mL
Cell Cycle and Replication
1031
Store at 4C in the dark.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.