Learn More
p19 INK4d Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00015APC
Product Code. 18681256
  • € 407.00 / 0.10mL
Description
p19 INK4d Polyclonal antibody specifically detects p19 INK4d in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D
Produced in rabbits immunized with purified, recombinant Human p19 INK4d (Accession#: P55273; Met10-Leu166)
Cell Cycle and Replication, Ovarian Carcinoma Cell Markers
Purified
Rabbit
0.1 mL
1032