missing translation for 'onlineSavingsMsg'
Learn More

p19ARF/CDKN2A Antibody (5-C3-1), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18659939 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18659939 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18659939 Supplier Novus Biologicals Supplier No. NB200174JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

p19ARF/CDKN2A Monoclonal antibody specifically detects p19ARF/CDKN2A in Human, Mouse, Golden Syrian Hamster (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF, Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen p19ARF/CDKN2A
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 5-C3-1
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias alternative reading frame, ARF, ARF-INK4a, CDK4I, CDKN2A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4), cyclin-dependent kinase inhibitor 2A, isoform 3, cyclin-dependent kinase inhibitor 2A, isoforms 1/2, cyclin-dependent kinase inhibitor protein, INK4, INK4a, Ink4a/Arf, INK4a-ARF, mitochondrial smARF', MTS1, p14, p14ARF, P16, p16(INK4a), p16-INK4, p16INK4a, p16-INK4a, P19, p19<ARF>, p19ARF, Pctr1
Host Species Rat
Immunogen A synthetic peptide containing amino acids 54-75 of the murine p19ARF/CDKN2A. [UniProt# Q64364]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 12578
Target Species Human, Mouse, Golden Syrian Hamster (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.