missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p21/CIP1/CDKN1A Antibody (CL13081), Novus Biologicals™
Mouse Monoclonal Antibody
€ 319.00 - € 526.00
Specifications
| Antigen | p21/CIP1/CDKN1A |
|---|---|
| Clone | CL13081 |
| Dilution | Western Blot 1 μg/ml, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681665
|
Novus Biologicals
NBP3-18554-25ul |
25 μg |
€ 319.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690885
|
Novus Biologicals
NBP3-18554-100ul |
100 μg |
€ 526.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p21/CIP1/CDKN1A Monoclonal antibody specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| p21/CIP1/CDKN1A | |
| Western Blot 1 μg/ml, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1 | |
| This antibody was developed against a sythetic peptide for P38936. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL13081 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific | |
| PBS, pH 7.2, 40% glycerol | |
| 1026 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title