missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCXR Antibody (OTI7D11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02192
This item is not returnable.
View return policy
Description
DCXR Monoclonal specifically detects DCXR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DCXR | |
| Monoclonal | |
| 0.51 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50 | |
| DCXR | |
| Full length human recombinant protein of human DCXR (NP_057370) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI7D11 | |
| Unconjugated | |
| Carbonyl reductase II, DCR, dicarbonyl/L-xylulose reductaseP34H, EC 1.1.1, EC 1.1.1.10, HCR2, HCRII, human carbonyl reductase 2, KIDCR, Kidney dicarbonyl reductase, L-xylulose reductase, SDR20C1, short chain dehydrogenase/reductase family 20C, member 1, Sperm surface protein P34H, XR | |
| Mouse | |
| 25.9 kDa | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Lipid and Metabolism | |
| 51181 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction