missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p23/PTGES3 Antibody (JJ3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-576
This item is not returnable.
View return policy
Description
p23/PTGES3 Monoclonal antibody specifically detects p23/PTGES3 in Human, Mouse, Rat, Chicken, Guinea Pig, Primate, Rabbit, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| p23/PTGES3 | |
| Monoclonal | |
| Unconjugated | |
| cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| p23 (JJ3) | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| JJ3 | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:1000 | |
| PTGES3 | |
| Recombinant human p23 protein. | |
| 100 μL | |
| Membrane Trafficking and Chaperones | |
| 10728 | |
| Human, Mouse, Rat, Chicken, Guinea Pig, Primate, Rabbit, Xenopus species | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction