missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p27/Kip1 Antibody (7B9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59444F
This item is not returnable.
View return policy
Description
p27/Kip1 Monoclonal antibody specifically detects p27/Kip1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| p27/Kip1 | |
| Monoclonal | |
| FITC | |
| CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| Recombinant human p27/Kip1 (1-198aa) purified from E. coli | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Prostate Cancer, Tumor Suppressors | |
| 1027 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 7B9 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction