missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p27/Kip1 Antibody (DCS-72.F6 + KIP1/769), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47771APC
This item is not returnable.
View return policy
Description
p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications
| p27/Kip1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| CDKN1B | |
| Mouse recombinant p27/Kip1 protein (DCS-72.F6); Recombinant human CDKN1B protein (KIP1/769) (Uniprot: P46527) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific, Prostate Cancer, Tumor Suppressors | |
| Recognizes a 27kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This monoclonal antibody co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| DCS-72.F6 + KIP1/769 | |
| Flow Cytometry | |
| CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction