missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p27/Kip1 Antibody (KIP1/769), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47769CL1
This item is not returnable.
View return policy
Description
p27/Kip1 Monoclonal antibody specifically detects p27/Kip1 in Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Flow (Intracellular),Immunofluorescence,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| p27/Kip1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| Recombinant full-length human p27/Kip1 protein (Uniprot: P46527) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific, Prostate Cancer, Tumor Suppressors | |
| 1027 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| KIP1/769 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction