Learn More
Invitrogen™ p27 Kip1 Recombinant Rabbit Monoclonal Antibody (23GB1825), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA554153
 View more versions of this product
Product Code. 30089831
  • € 217.00 / 20µL
Description
p27 Kip1 functions as a negative regulator of G1 mouse expression and has been proposed to function as a possible mediator of TGFb induced G1 arrest. p27 Kip1 is an important regulator of cell cycle progression, a potent inhibitor of cyclin E- and cyclin A-CDK2 complexes. p27 Kip1 forms a complex with cyclin type D-CDK4 complexes and is involved in the assembly, stability, and modulation of CCND1-CDK4 complex activation. p27 Kip1 acts either as an inhibitor or an activator of cyclin type D-CDK4 complexes depending on its phosphorylation state and/or stoichometry. p27 Kip1 has a molecular weight of approximately 27kDa and the degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p27 Kip1
Recombinant Monoclonal
Unconjugated
Cdkn1b
AA408329; AI843786; Cdki1b; CDKN1B; CDKN4; CDN1B; cyclin dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor 1B (p27 Kip1); Cyclin-dependent kinase inhibitor 1B (p27, Kip1); cyclin-dependent kinase inhibitor p27; Cyclin-dependent kinase inhibitor p27 Kip1; cyclin-dependent kinase inhibitor p27/Kip1; KIP1; MEN1B; MEN4; p27; P27 CDKN1B; p27(KIP1); p27Kip1
Rabbit
Affinity chromatography
RUO
1027, 83571
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB1825
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P46527
Cdkn1b
A synthesized peptide derived from human p27 KIP 1 (150-198AA).
20 μL
Primary
Human, Rat