missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2RY1 Mouse anti-Human, Clone:4G5D6, Abnova™
Mouse monoclonal antibody raised against recombinant human P2RY1.
Brand: Abnova MAB17957.100ug
This item is not returnable.
View return policy
Description
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor functions as a receptor for extracellular ATP and ADP. In platelets binding to ADP leads to mobilization of intracellular calcium ions via activation of phospholipase C, a change in platelet shape, and probably to platelet aggregation. [provided by RefSeq]Specifications
| P2RY1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| P2RY1 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Western Blot | |
| 4G5D6 | |
| purinergic receptor P2Y, G-protein coupled, 1 | |
| In PBS (0.05% sodium azide) | |
| P2Y1 | |
| Recombinant protein corresponding to human P2RY1. | |
| 5028 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction