missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P2X4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541080
This item is not returnable.
View return policy
Description
Peptide sequence: VQLLILAYVI GWVFVWEKGY QETDSVVSSV TTKVKGVAVT NTSKLGFRIW Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 86%.
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel with high calcium permeability. The main pharmacological distinction between the members of the purinoceptor family is the relative sensitivity to the antagonists suramin and PPADS. The product of this gene has the lowest sensitivity for these antagonists. Multiple alternatively spliced transcript variants have been identified for this gene although their full-length natures have not been determined.
Specifications
| P2X4 | |
| Polyclonal | |
| Unconjugated | |
| P2RX4 | |
| AI504491; ATP receptor; ATP-gated cation channel protein; AW555605; D5Ertd444e; ionotropic purinergic receptor; P2 purinergic receptor subtype; P2RX4; P2X purinoceptor 4; P2X receptor, subunit 4; P2X4; P2X4 purinoceptor; P2x4 receptor; P2X4R; Purinergic receptor; purinergic receptor P2X 4; purinergic receptor P2X, ligand gated ion channel, 4; purinergic receptor P2X, ligand-gated ion channel 4; purinergic receptor P2X, ligand-gated ion channel, 4; purinergic receptor P2X4; purinergic receptor P2X4 isoform a; purinergic receptor P2X4 subunit variant 1; purinergic receptor P2X4 subunit variant 2; purinergic receptor P2x4 subunit variant e; purinoceptor P2X4; putative partial extracellular loop region | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5025 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99571 | |
| P2RX4 | |
| Synthetic peptide directed towards the N-terminal of human P2RX4 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction