missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X5/P2RX5 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98199AF647
This item is not returnable.
View return policy
Description
P2X5/P2RX5 Polyclonal specifically detects P2X5/P2RX5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| P2X5/P2RX5 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| ATP receptor, ATP receptor subunit, ionotropic ATP receptor P2X5, LRH-1, MGC47755, P2X purinoceptor 5, P2X5lymphoid-restricted histocompatibility antigen-1, P2X5R, Purinergic receptor, purinergic receptor P2X ligand gated ion channel 5, purinergic receptor P2X, ligand-gated ion channel, 5 | |
| Produced in rabbits immunized with E. coli-derived Human P2X5/P2RX5 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 5026 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction