missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P2X7 Monoclonal Antibody (7G1D6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531813
This item is not returnable.
View return policy
Description
MA5-31813 has been tested in indirect ELISA.
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and is responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Activation of this nuclear receptor by ATP in the cytoplasm may be a mechanism by which cellular activity can be coupled to changes in gene expression. Multiple alternatively spliced variants have been identified, most of which fit nonsense-mediated decay criteria.
Specifications
| P2X7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q99572, Q9Z1M0 | |
| P2RX7 | |
| Purified recombinant fragment of human P2RX7 (AA: 226-452) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 7G1D6 | |
| Unconjugated | |
| P2RX7 | |
| AI467586; ATP receptor; MGC20089; P2rx7; P2X purinoceptor 7; p2X receptor 7; P2X(7); P2X7; P2X7 purinoceptor; P2X7 receptor; P2X7R; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X 7; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7; purinergic receptor P2X7 variant A | |
| Mouse | |
| Protein G | |
| RUO | |
| 18439, 5027 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction