missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7/P2RX7 Antibody (1058613), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB97903MFV500
This item is not returnable.
View return policy
Description
P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Flow Cytometry
Specifications
| P2X7/P2RX7 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1058613 | |
| Flow Cytometry | |
| ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| Human P2X7/P2RX7 synthetic peptide, Accession # Q99572 | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 5027 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction