missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y1/P2RY1 Antibody (4G5D6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 217.00 - € 582.00
Spécification
| Antigen | P2Y1/P2RY1 |
|---|---|
| Clone | 4G5D6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
18267442
|
Novus Biologicals
NBP2-61664 |
0.1 mL |
€ 582.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631118
|
Novus Biologicals
NBP2-61664-0.025ml |
0.025 mL |
€ 217.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2Y1/P2RY1 Monoclonal specifically detects P2Y1/P2RY1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Spécification
| P2Y1/P2RY1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| 5028 | |
| Purified recombinant fragment of human P2RY1 (AA: extra mix) expressed in E. Coli. | |
| Primary | |
| 42 kDa |
| 4G5D6 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| ATP receptor, P2 purinoceptor subtype Y1, P2Y purinoceptor 1, P2Y1platelet ADP receptor, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 1 | |
| P2RY1 | |
| IgG2b | |
| Protein G purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit