missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y12/P2RY12 Antibody (1C2A9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-61749
This item is not returnable.
View return policy
Description
P2Y12/P2RY12 Monoclonal specifically detects P2Y12/P2RY12 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| P2Y12/P2RY12 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999 | |
| Mouse | |
| 39.4 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 1C2A9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| P2RY12 | |
| Purified recombinant fragment of human P2RY12 (AA: extra mix) expressed in E. Coli. | |
| Protein G purified | |
| GPCR | |
| 64805 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction