Learn More
Invitrogen™ P2Y4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534090
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Monkey, Panda, Rabbit (94%); Bovine, Opossum (88%); Mouse, Rat, Bat, Pig (81%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Receptor for UTP and UDP coupled to G-proteins that activate a phosphatidylinositol-calcium second messenger system. Not activated by ATP or ADP.
Specifications
| P2Y4 | |
| Polyclonal | |
| Unconjugated | |
| P2RY4 | |
| NRU; P2P; P2RY4; P2Y ATP receptor 4; P2Y purinoceptor 4; P2Y4; P2Y4 receptor; P2Y4R; P2YR4; purinergic receptor P2Y G-protein coupled 4; purinergic receptor P2Y, G-protein coupled 4; purinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y4; RP13-26D14.5; UNR; Uridine nucleotide receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5030 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P51582 | |
| P2RY4 | |
| Synthetic 16 amino acid peptide from internal region of human P2RY4 / P2Y4. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.