missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P2Y4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550523
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total P2Y4 protein.
Receptor for UTP and UDP coupled to G-proteins that activate a phosphatidylinositol-calcium second messenger system. Not activated by ATP or ADP.
Specifications
| P2Y4 | |
| Polyclonal | |
| Unconjugated | |
| P2RY4 | |
| NRU; P2P; P2RY4; P2Y ATP receptor 4; P2Y purinoceptor 4; P2Y4; P2Y4 receptor; P2Y4R; P2YR4; purinergic receptor P2Y G-protein coupled 4; purinergic receptor P2Y, G-protein coupled 4; purinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y4; RP13-26D14.5; UNR; Uridine nucleotide receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5030 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P51582 | |
| P2RY4 | |
| Synthetic peptide corresponding to residues near the C terminal of human pyrimidinergic receptor P2Y, G-protein coupled, 4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction