missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y6/P2RY6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS2726
This item is not returnable.
View return policy
Description
P2Y6/P2RY6 Polyclonal antibody specifically detects P2Y6/P2RY6 in Human, Rat, Porcine, Bovine, Hamster, Primate, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| P2Y6/P2RY6 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| G-coupled nucleotide receptor, MGC15335, P2 purinoceptor, P2Y6 receptor, P2Y6P2Y purinoceptor 6, pyrimidinergic receptor P2Y, G-protein coupled, 6 | |
| Synthetic 17 amino acid peptide from 2nd cytoplasmic domain of human P2Y6/P2RY6. | |
| 0.05 mL | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| 5031 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 17 - 34 μg/mL | |
| Q15077 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Bovine, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction