missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y6/P2RY6 Rabbit anti-Human, Clone: 4I8C9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16259-20UL
This item is not returnable.
View return policy
Description
P2Y6/P2RY6 Monoclonal antibody specifically detects P2Y6/P2RY6 in Human samples. It is validated for Western Blot
Specifications
| P2Y6/P2RY6 | |
| Monoclonal | |
| Unconjugated | |
| G-coupled nucleotide receptor, MGC15335, P2 purinoceptor, P2Y6 receptor, P2Y6P2Y purinoceptor 6, pyrimidinergic receptor P2Y, G-protein coupled, 6 | |
| A synthetic peptide corresponding to a sequence within amino acids 229-328 of human P2Y6/P2RY6 (Q15077). | |
| 20 μg | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| 5031 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 4I8C9 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction