missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y8 Antibody (1G5A2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61765
This item is not returnable.
View return policy
Description
P2Y8 Monoclonal specifically detects P2Y8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| P2Y8 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| G-protein coupled purinergic receptor P2Y8, P2Y purinoceptor 8, P2Y8MGC50878, purinergic receptor P2Y, G-protein coupled, 8 | |
| Mouse | |
| 40.6 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1G5A2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| P2RY8 | |
| Purified recombinant fragment of human P2RY8 (AA: extra mix) expressed in E. Coli. | |
| Protein G purified | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| 286530 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction