missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 alpha Antibody (9F12), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-96875FR
This item is not returnable.
View return policy
Description
p38 alpha Monoclonal specifically detects p38 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.
Specifications
| p38 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A | |
| Mouse | |
| Protein G purified | |
| Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
| 1432 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 9F12 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry | |
| NP_001306.1 | |
| MAPK14 | |
| Full length recombinant protein expressed in E.coli cells | |
| 0.1 mL | |
| Primary | |
| Detects approx 38kDa. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction