missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 delta/SAPK4 Antibody (OTI12B2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73188DL594
This item is not returnable.
View return policy
Description
p38 delta/SAPK4 Monoclonal antibody specifically detects p38 delta/SAPK4 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
| p38 delta/SAPK4 | |
| Monoclonal | |
| DyLight 594 | |
| EC 2.7.11, EC 2.7.11.24, MAP kinase 13, MAPK 13, MGC99536, mitogen-activated protein kinase 13, Mitogen-activated protein kinase p38 delta, p38delta, PRKM13, SAPK4MAP kinase p38 delta, Stress-activated protein kinase 4 | |
| Full length recombinant protein of human p38 delta/SAPK4 (NP_002745) produced in HEK293T cells. | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 5603 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| OTI12B2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction