missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p40 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99500AF594
This item is not returnable.
View return policy
Description
p40 Polyclonal antibody specifically detects p40 in Human samples. It is validated for Western Blot
Specifications
| p40 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 40 kDa Rab9 effector protein, bA65N13.1, p40, Rab9 effector p40, Rab9 effector protein with kelch motifs, RAB9P40DKFZp686P1077, RABEPK | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human p40. | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 10244 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction