missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p40/deltaNp63 Antibody (P40/8225R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20793AF700
This item is not returnable.
View return policy
Description
p40/deltaNp63 Monoclonal antibody specifically detects p40/deltaNp63 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| p40/deltaNp63 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| AIS, amplified in squamous cell carcinoma, B(p51A), B(p51B), Chronic μgcerative stomatitis protein, EEC3, Keratinocyte transcription factor KET, KET, LMS, OFC8, p53CP, SHFM4, TP53CP, TP53L, TP63, TP73L, Transformation-related protein 63, tumor protein p53-competing protein | |
| Recombinant fragment (around aa1-200) of human p40/deltaNp63 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| P40/8225R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8626 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction