missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p40 Recombinant Rabbit Monoclonal Antibody (24GB1540)

Rabbit Recombinant Monoclonal Antibody
€ 198.00 - € 564.00
Specifications
| Antigen | p40 |
|---|---|
| Clone | 24GB1540 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276240
![]() |
Invitrogen™
MA557668 |
100 μL |
€ 564.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275295
|
Invitrogen™
MA557669 |
20 μL |
€ 198.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p40 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.Specifications
| p40 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Rabepk | |
| 40 kDa Rab9 effector protein; 8430412M01Rik; 9530020d24rik; AV073337; bA65N13.1; C87311; p40; Rab9 effector p40; rab9 effector protein with kelch motifs; RAB9P40; RABEPK; RGD1310612 | |
| Rabepk | |
| Primary | |
| Affinity Chromatography |
| 24GB1540 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7Z6M1 | |
| 10244 | |
| A synthesized peptide derived from human p40 (aa 250-372). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title