missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p400 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-88230-25ul
This item is not returnable.
View return policy
Description
p400 Polyclonal specifically detects p400 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| p400 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CAG repeat protein 32, CAGH32hDomino, DKFZP434I225, Domino homolog, E1A binding protein p400, E1A-binding protein p400, EC 3.6.1, EC 3.6.4.-, FLJ45115, KIAA1498p400 kDa SWI2/SNF2-related protein, KIAA1818, P400, p400 SWI2/SNF2-related protein, TNRC12FLJ42018, trinucleotide repeat containing 12, Trinucleotide repeat-containing gene 12 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EP400 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DDEVDDEEETIEEEEANEGVVDHQTELSNLAKEAELPLLDLMKLYEGAFLPSSQWPRPKPDGEDTSGEEDADDCPGDRESRKDLVLIDSLFIMDQFKAAERMNIGKPNAKDIADVTAVAEAILPKGSARVTTSVKFNAPSL | |
| 25 μL | |
| Cancer, Tumor Suppressors | |
| 57634 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction