missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p41 Flagellin Antibody (6802), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08849AF488
This item is not returnable.
View return policy
Description
p41 Flagellin Monoclonal antibody specifically detects p41 Flagellin in Bacteria samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| p41 Flagellin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 41 kDa antigen, fla, Flagellar filament 41 kDa core protein, Flagellin, p41 | |
| A crude sonicate of Borrelia spirochetes | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Purified |
| ELISA, Immunofluorescence | |
| 6802 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 46847386 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction