missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P4HA3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA548623

Product Code. 15602925

  • € 520.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

P4HA3 Polyclonal Antibody for Western Blot

Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P4HA3
Polyclonal
Unconjugated
P4ha3
4-PH alpha-3; collagen prolyl 4-hydroxylase alpha 3; collagen prolyl 4-hydroxylase alpha III subunit; collagen prolyl 4-hydroxylase alpha(III); C-P4H alpha III; D930031A02Rik; P4ha3; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide III; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-3; prolyl 4-hydroxylase subunit alpha 3; prolyl 4-hydroxylase subunit alpha-3; prolyl 4-hydroxylase, alpha polypeptide III; UNQ711/PRO1374
Rabbit
Antigen affinity chromatography, Protein A
RUO
283208
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.3 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q7Z4N8
P4ha3
KLH conjugated synthetic peptide between 274-303 amino acids from the Central region of human P4HA3.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.