missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P4HA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552483
This item is not returnable.
View return policy
Description
Immunogen sequence: TTPVANPLLA FTLIKRLQSD WRNVVHSLEA SENIRALKDG YEKVEQDLPA FEDLEGAARA LMRLQDVYML NVKGLARGVF QRVTGSAITD LYSPKRLFSL TGDDCFQVGK V Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.
Specifications
| P4HA3 | |
| Polyclonal | |
| Unconjugated | |
| P4ha3 | |
| 4-PH alpha-3; collagen prolyl 4-hydroxylase alpha 3; collagen prolyl 4-hydroxylase alpha III subunit; collagen prolyl 4-hydroxylase alpha(III); C-P4H alpha III; D930031A02Rik; P4ha3; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide III; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-3; prolyl 4-hydroxylase subunit alpha 3; prolyl 4-hydroxylase subunit alpha-3; prolyl 4-hydroxylase, alpha polypeptide III; UNQ711/PRO1374 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283208 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z4N8 | |
| P4ha3 | |
| Recombinant protein corresponding to Human P4HA3. Recombinant protein control fragment (Product #RP-89693). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction