Learn More
p53 Antibody (CL2199), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-34495
 View more versions of this product
Product Code. 18790714
  • € 572.00 / 0.10mL
Description
p53 Monoclonal specifically detects p53 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated, PCR.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
IgG1
Western Blot 1 μg/mL, Simple Western 1:50, Immunohistochemistry 1:200 - 1:5000, Immunocytochemistry/Immunofluorescence 2 to 10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, KnockDown Validated
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.