missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Antibody (TP53/3890R), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21044JF549
This item is not returnable.
View return policy
Description
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| p53 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
| Recombinant human full-length p53 protein | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 7157 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| TP53/3890R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction