missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Antibody (V5P4H6*B3), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50231AF594
This item is not returnable.
View return policy
Description
p53 Monoclonal specifically detects p53 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| p53 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TP53 | |
| Ovalbumin-conjugated synthetic peptide KLMVKDEGRS (C-terminal sequence) | |
| 0.1 mL | |
| Primary | |
| Zebrafish | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| V5P4H6*B3 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
| Mouse | |
| Protein A purified | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 7157.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction