missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p57 Kip2 Antibody (KIP2/7187) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20895JF669
This item is not returnable.
View return policy
Description
p57 Kip2 Monoclonal antibody specifically detects p57 Kip2 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| p57 Kip2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Beckwith-Wiedemann syndrome, BWS, cyclin-dependent kinase inhibitor 1C, cyclin-dependent kinase inhibitor 1C (p57, Kip2), Cyclin-dependent kinase inhibitor p57, KIP2BWCR, p57, p57Kip2, WBS | |
| Recombinant full-length human p57 Kip2 protein | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 1028 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| KIP2/7187 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction