Learn More
p62/SQSTM1 Antibody (864807), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8028MFV610
Product Code. 30065397
  • € 719.00 / 0.10mL
Description
p62/SQSTM1 Monoclonal antibody specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p62/SQSTM1
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
864807
Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Knockout Validated
A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
E. coli-derived recombinant human p62/SQSTM1, Asp368-Leu440, Accession # Q13501
0.1 mL
Adaptive Immunity, Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics, Innate Immunity, Tumor Suppressors
8878
Store at 4C in the dark.