missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p62/SQSTM1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-48320PE
This item is not returnable.
View return policy
Description
p62/SQSTM1 Polyclonal antibody specifically detects p62/SQSTM1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating
Specifications
| p62/SQSTM1 | |
| Polyclonal | |
| PE | |
| A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3 | |
| Partial recombinant protein made to the C terminal of the human p62/SQSTM1 protein (within residues 300-440). [Swiss-Prot Q13501] | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics, Innate Immunity, Tumor Suppressors | |
| 8878 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 1.0 mg/mL | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction