missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63/TP73L Antibody (I27-I), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08205-200ul
This item is not returnable.
View return policy
Description
p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| p63/TP73L | |
| Monoclonal | |
| Unconjugated | |
| 20 mM Tris-HCl, pH 8.0, 20 mg/ml BSA | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| I27-I | |
| Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin 1:100-1:200 | |
| amplified in squamous cell carcinoma, Chronic μgcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
| Peptide derived from the internal sequence of human p63/TP73L. Antibody recognizes the epitope between Arg360 - His383. | |
| 200 μg | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors | |
| 8626 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction