missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63/TP73L Antibody (TP63/1786), Novus Biologicals™
Mouse Monoclonal Antibody
€ 249.00 - € 484.00
Specifications
| Antigen | p63/TP73L |
|---|---|
| Clone | TP63/1786 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA, Protein Array |
| Applications | Western Blot, ELISA, Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661877
|
Novus Biologicals
NBP3-07441-20ug |
20 μg |
€ 249.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613127
|
Novus Biologicals
NBP3-07441-100ug |
100 μg |
€ 484.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human samples. It is validated for Western Blot, ELISA, Protein ArraySpecifications
| p63/TP73L | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Protein Array | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| amplified in squamous cell carcinoma, Chronic μgcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
| Recombinant fragment (around aa 3-106) of human p63/TP73L protein (exact sequence is proprietary) (Uniprot: Q9H3D4) | |
| Primary | |
| Store at 4°C. |
| TP63/1786 | |
| Western Blot 1-2 μg/mL, ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q9H3D4, Q9H3D4 | |
| 8626 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title