missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63/TP73L Antibody (TP63/2428), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07443-100ug
This item is not returnable.
View return policy
Description
p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| p63/TP73L | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Q9H3D4, Q9H3D4 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin), Protein Array | |
| TP63/2428 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| amplified in squamous cell carcinoma, Chronic μgcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
| Recombinant full-length human p63/TP73L protein (Uniprot: Q9H3D4 ) | |
| 100 μg | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors | |
| 8626 | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction