missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63/TP73L Antibody (ZR8), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 309.00 - € 551.00
Specifications
| Antigen | p63/TP73L |
|---|---|
| Clone | ZR8 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18675076
|
Novus Biologicals
NBP3-07454-100ug |
100 μg |
€ 551.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672889
|
Novus Biologicals
NBP3-07454-20ug |
20 μg |
€ 309.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| p63/TP73L | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| amplified in squamous cell carcinoma, Chronic μgcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
| A synthetic peptide (ENNAQTQFSEPQY) corresponding to aa5-17 of human p63/TP73L protein (Uniprot: Q9H3D4 ) | |
| Primary | |
| Store at 4°C. |
| ZR8 | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q9H3D4, Q9H3D4 | |
| 8626 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title