missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p67phox/NOXA2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97097AF488
This item is not returnable.
View return policy
Description
p67phox/NOXA2 Polyclonal specifically detects p67phox/NOXA2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| p67phox/NOXA2 | |
| Polyclonal | |
| Western Blot, ELISA | |
| autosomal 2, NADPH oxidase activator 2, NCF-2, neutrophil cytosol factor 2, neutrophil cytosolic factor 2, neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal2), NOXA2FLJ93058, p67phox, p67-phox | |
| Produced in rabbits immunized with purified, recombinant p67phox/NOXA2 (AAH01606.1; Met1-Val526). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4688 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction