missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p70 S6 Kinase beta/S6K2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-1545
This item is not returnable.
View return policy
Description
p70 S6 Kinase beta/S6K2 Polyclonal specifically detects p70 S6 Kinase beta/S6K2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| p70 S6 Kinase beta/S6K2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, KLS, p70 ribosomal S6 kinase beta, p70 S6 kinase beta, p70 S6KB, p70 S6K-beta, p70(S6K)-beta, p70-beta, P70-beta-1, P70-beta-2, p70-S6K 2, P70S6K2, p70S6Kb, ribosomal protein S6 kinase beta-2, ribosomal protein S6 kinase, 70kD, polypeptide 2,70 kDa ribosomal protein S6 kinase 2, ribosomal protein S6 kinase, 70kDa, polypeptide 2, S6K2, S6K-beta, S6K-beta2, S6K-beta-2, serine/threonine-protein kinase 14 beta, Serine/threonine-protein kinase 14B, SRK, STK14BS6 kinase-related kinase | |
| Rabbit | |
| 53 kDa | |
| 0.1 mL | |
| Autophagy, Cancer, Cell Cycle and Replication | |
| 6199 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2 -6 ug/mg of lysate | |
| Q9UBS0 | |
| RPS6KB2 | |
| The immunogen recognized by this antibody maps to a region between residue 430 and the C-terminus (residue 482) of human Ribosomal Protein S6 Kinase, 70kda, polypeptide 2 using the numbering given in entry NP_003943.2 (GeneID 6199). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Chimpanzee, Naked mole rat, European domestic ferret, Northern white-cheeked gibbon, and Thirteen-lined ground squirrel. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction